Assay Details
Gene Symbol: GDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034686.2 | NP_001029858.1 | 1 - 2 | 330 | 93 | |
GenBank mRNA | BC102273.1 | - | 1 - 2 | 331 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
