Assay Details
Gene Symbol: RAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034705.2 | NP_001029877.1 | 1 - 2 | 34 | 112 | |
GenBank mRNA | BC102559.1 | - | 1 - 2 | 35 | 112 | |
BC151426.1 | - | 1 - 2 | 40 | 112 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102559.1, BC151426.1, NM_001034705.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
