Assay Details
Gene Symbol: RAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034705.2 | NP_001029877.1 | 5 - 6 | 488 | 88 | |
GenBank mRNA | BC102559.1 | - | 5 - 6 | 489 | 88 | |
BC151426.1 | - | 5 - 6 | 494 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
