Assay Details
Gene Symbol: TRAPPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034796.1 | NP_001029968.1 | 1 - 2 | 61 | 71 | |
GenBank mRNA | BC102418.1 | - | 1 - 2 | 61 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102418.1, NM_001034796.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
