Assay Details
Gene Symbol: LOC616948 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034798.1 | NP_001029970.1 | 2 - 3 | 633 | 105 | |
GenBank mRNA | AY970972.1 | - | 2 - 3 | 633 | 105 | |
DQ381151.1 | - | 1 - 2 | 422 | 105 |
Gene Symbol: TRIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205305.1 | NP_001192234.1 | 2 - 3 | 800 | 105 | |
GenBank mRNA | BC119899.1 | - | 2 - 3 | 800 | 105 | |
DQ380509.1 | - | 1 - 2 | 422 | 105 | ||
DQ381150.1 | - | 1 - 2 | 422 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
