Assay Details
Gene Symbol: PTGR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035281.2 | NP_001030358.1 | 5 - 6 | 417 | 83 | |
GenBank mRNA | BC102822.1 | - | 5 - 6 | 419 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
