Assay Details
Gene Symbol: RPL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035306.2 | NP_001030383.1 | 1 | 400 | 109 | |
GenBank mRNA | BC105198.1 | - | 1 | 401 | 109 | |
BT021482.1 | - | 4 - 5 | 377 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
