Assay Details
Gene Symbol: RPL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035306.2 | NP_001030383.1 | 1 | 777 | 130 | |
GenBank mRNA | BC105198.1 | - | 1 | 778 | 130 | |
BT021482.1 | - | 6 - 7 | 754 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
