Assay Details
Gene Symbol: NEDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035333.2 | NP_001030410.1 | 14 - 15 | 1934 | 150 | |
GenBank mRNA | BC107541.1 | - | 14 - 15 | 1935 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
