Assay Details
Gene Symbol: OXA1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035335.2 | NP_001030412.1 | 9 - 10 | 1191 | 61 | |
GenBank mRNA | BC103367.1 | - | 9 - 10 | 1194 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
