Assay Details
Gene Symbol: ARID5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035349.1 | NP_001030426.1 | 3 - 4 | 314 | 82 | |
GenBank mRNA | BC104604.1 | - | 3 - 4 | 314 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
