Assay Details
Gene Symbol: SC5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035356.1 | NP_001030433.1 | 3 - 4 | 420 | 115 | |
GenBank mRNA | BC103338.1 | - | 3 - 4 | 420 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
