Assay Details
Gene Symbol: RPL28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035503.2 | NP_001030580.1 | 1 - 2 | 40 | 72 | |
GenBank mRNA | BC102344.1 | - | 1 - 2 | 41 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102344.1, NM_001035503.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
