Assay Details
Gene Symbol: RPL28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035503.2 | NP_001030580.1 | 4 - 5 | 367 | 101 | |
GenBank mRNA | BC102344.1 | - | 4 - 5 | 368 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
