Assay Details
Gene Symbol: C1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037448.2 | NP_001032525.1 | 2 - 3 | 193 | 140 | |
GenBank mRNA | BC108147.1 | - | 2 - 3 | 194 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
