Assay Details
Gene Symbol: FCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037452.1 | NP_001032529.1 | 7 - 8 | 577 | 117 | |
GenBank mRNA | BC108165.1 | - | 7 - 8 | 577 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
