Assay Details
Gene Symbol: EEF1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037464.2 | NP_001032541.1 | 3 - 4 | 428 | 75 | |
GenBank mRNA | BC108110.1 | - | 3 - 4 | 429 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
