Assay Details
Gene Symbol: FKBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037481.1 | NP_001032558.1 | 3 - 4 | 339 | 101 | |
NM_001113256.1 | NP_001106727.1 | 4 - 5 | 442 | 101 | ||
GenBank mRNA | BC108190.1 | - | 3 - 4 | 339 | 101 | |
BI849286.1 | - | 4 - 5 | 428 | 101 | ||
CN433599.1 | - | 4 - 5 | 362 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
