Assay Details
Gene Symbol: SLIRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037485.2 | NP_001032562.1 | 2 - 3 | 158 | 96 | |
GenBank mRNA | BC108247.1 | - | 2 - 3 | 160 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
