Assay Details
Gene Symbol: MRPL42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037622.2 | NP_001032711.1 | 5 - 6 | 437 | 151 | |
GenBank mRNA | BC109670.1 | - | 5 - 6 | 438 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
