Assay Details
Gene Symbol: TNFRSF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040490.2 | NP_001035580.1 | 3 - 4 | 380 | 66 | |
GenBank mRNA | BC105222.1 | - | 3 - 4 | 381 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
