Assay Details
Gene Symbol: TNFRSF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040490.2 | NP_001035580.1 | 7 - 8 | 940 | 71 | |
GenBank mRNA | BC105222.1 | - | 7 - 8 | 941 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
