Assay Details
Gene Symbol: TNFRSF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040490.2 | NP_001035580.1 | 9 - 10 | 1180 | 74 | |
GenBank mRNA | AF031589.1 | - | 1 - 2 | 113 | 74 | |
BC105222.1 | - | 9 - 10 | 1181 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
