Assay Details
Gene Symbol: GLRX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040523.2 | NP_001035613.1 | 4 - 5 | 572 | 131 | |
GenBank mRNA | BC109742.1 | - | 4 - 5 | 573 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
