Assay Details
Gene Symbol: UOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075648.2 | NP_001069116.1 | 6 - 7 | 802 | 82 | |
GenBank mRNA | BC105244.1 | - | 6 - 7 | 803 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
