Assay Details
Gene Symbol: ROPN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075717.2 | NP_001069185.1 | 4 - 5 | 806 | 86 | |
GenBank mRNA | BC102601.1 | - | 4 - 5 | 808 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
