Assay Details
Gene Symbol: COMMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076999.1 | NP_001070467.1 | 7 - 8 | 540 | 96 | |
GenBank mRNA | BC103075.1 | - | 7 - 8 | 540 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
