Assay Details
Gene Symbol: RPS14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077830.2 | NP_001071298.1 | 1 - 2 | 36 | 81 | |
GenBank mRNA | BC102536.1 | - | 1 - 2 | 37 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102536.1, NM_001077830.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
