Assay Details
Gene Symbol: CSRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083771.1 | NP_001077240.1 | 3 - 4 | 329 | 77 | |
GenBank mRNA | BC104539.2 | - | 3 - 4 | 329 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
