Assay Details
Gene Symbol: MDK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173935.3 | NP_776360.2 | 4 - 5 | 454 | 86 | |
GenBank mRNA | BC103211.1 | - | 4 - 5 | 455 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
