Assay Details
Gene Symbol: ENO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174049.2 | NP_776474.2 | 1 - 2 | 135 | 82 | |
GenBank mRNA | BC103354.1 | - | 1 - 2 | 135 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC103354.1, NM_174049.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
