Assay Details
Gene Symbol: NEDD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174764.3 | NP_777189.1 | 1 - 2 | 90 | 80 | |
GenBank mRNA | AF227256.1 | - | 1 - 2 | 55 | 80 | |
BC102306.1 | - | 1 - 2 | 90 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102306.1, NM_174764.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
