Assay Details
Gene Symbol: NDUFB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176656.4 | NP_788829.1 | 4 - 5 | 343 | 66 | |
GenBank mRNA | AY911319.1 | - | 4 - 5 | 347 | 66 | |
BC102690.1 | - | 4 - 5 | 344 | 66 | ||
X63208.1 | - | 350 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
