Assay Details
Gene Symbol: RASSF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024549.2 | NP_001019720.1 | 1 - 2 | 86 | 102 | |
GenBank mRNA | BC111208.1 | - | 1 - 2 | 86 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC111208.1, NM_001024549.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
