Assay Details
Gene Symbol: CHMP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191260.2 | NP_001178189.1 | 1 - 2 | 135 | 85 | |
GenBank mRNA | BC109956.1 | - | 1 - 2 | 136 | 85 | |
DT837819.1 | - | 1 - 2 | 136 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC109956.1, NM_001191260.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
