Assay Details
Gene Symbol: CHMP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191260.2 | NP_001178189.1 | 4 - 5 | 380 | 58 | |
GenBank mRNA | BC109956.1 | - | 4 - 5 | 381 | 58 | |
DT837819.1 | - | 4 - 5 | 381 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
