Assay Details
Gene Symbol: GOT1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038058.1 | NP_001033147.1 | 5 - 6 | 723 | 93 | |
GenBank mRNA | BC111285.1 | - | 5 - 6 | 723 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
