Assay Details
Gene Symbol: POLR2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038093.2 | NP_001033182.1 | 6 - 7 | 636 | 56 | |
GenBank mRNA | BC111280.1 | - | 6 - 7 | 638 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
