Assay Details
Gene Symbol: METTL26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038106.2 | NP_001033195.1 | 1 - 2 | 380 | 91 | |
GenBank mRNA | BC109870.1 | - | 1 - 2 | 382 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
