Assay Details
Gene Symbol: REXO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038154.2 | NP_001033243.1 | 8 - 9 | 1216 | 107 | |
GenBank mRNA | BC111262.1 | - | 8 - 9 | 1217 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
