Assay Details
Gene Symbol: CNN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038179.1 | NP_001033268.1 | 1 | 480 | 126 | |
GenBank mRNA | BC109697.1 | - | 1 | 480 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
