Assay Details
Gene Symbol: PSMB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038539.2 | NP_001033628.1 | 5 - 6 | 583 | 78 | |
GenBank mRNA | BC109494.1 | - | 5 - 6 | 584 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
