Assay Details
Gene Symbol: GAMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038544.2 | NP_001033633.1 | 4 - 5 | 496 | 91 | |
GenBank mRNA | BC109825.1 | - | 4 - 5 | 497 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
