Assay Details
Gene Symbol: RRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038688.2 | NP_001033777.1 | 5 - 6 | 589 | 67 | |
GenBank mRNA | BC110229.1 | - | 5 - 6 | 590 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
