Assay Details
Gene Symbol: RPL7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040520.2 | NP_001035610.1 | 6 - 7 | 655 | 66 | |
GenBank mRNA | BC109810.1 | - | 6 - 7 | 656 | 66 | |
BT030549.1 | - | 5 - 6 | 631 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
