Assay Details
Gene Symbol: BAG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040544.2 | NP_001035634.1 | 1 - 2 | 205 | 78 | |
GenBank mRNA | BC111177.1 | - | 1 - 2 | 206 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC111177.1, NM_001040544.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
