Assay Details
Gene Symbol: PRSS37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040569.2 | NP_001035659.1 | 2 - 3 | 134 | 98 | |
GenBank mRNA | BC109928.1 | - | 2 - 3 | 135 | 98 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC109928.1, NM_001040569.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
