Assay Details
Gene Symbol: HAX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045953.2 | NP_001039418.2 | 1 - 2 | 152 | 114 | |
GenBank mRNA | BC112670.1 | - | 1 - 2 | 108 | 114 | |
EV657492.1 | - | 1 - 2 | 159 | 114 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC112670.1, NM_001045953.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
