Assay Details
Gene Symbol: DIMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046036.2 | NP_001039501.1 | 3 - 4 | 292 | 86 | |
GenBank mRNA | BC112886.1 | - | 3 - 4 | 293 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
