Assay Details
Gene Symbol: DIMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046036.2 | NP_001039501.1 | 9 - 10 | 786 | 116 | |
GenBank mRNA | BC112886.1 | - | 9 - 10 | 787 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
